Learn More
M&I Materials Apiezon™ H Grease
Low vapor pressure sealing agents and lubricants for high-vacuum applications
Supplier:  M&I Materials FAPGH00025
Catalog No. 14-638-15A
Only null left
Explore available promotions
Description
Description
Low vapor pressure sealing agents and lubricants for high-vacuum applications
Apiezon products are silicone-free, high-purity hydrocarbons distilled from selected petroleum feedstock in a molecular still.
  • H Grease has the broadest range
Specifications
Specifications
H Grease
1.7 x 109 torr
14°F to 464°F
0.881 oz.
Low vapor pressure sealing agents and lubricants for high-vacuum applications
10 Pk.
-10°C to +240°C
25 mL
Product Suggestions
Product Suggestions