missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Aphidicolin, ACROS Organics™
$671.56 - $671.56
Chemical Identifiers
CAS | 38966-21-1 |
---|---|
Molecular Formula | C20H34O4 |
Molecular Weight (g/mol) | 338.488 |
MDL Number | MFCD00083214 |
InChI Key | NOFOAYPPHIUXJR-APNQCZIXSA-N |
Synonym | +-aphidicolin, +/--aphidicolin, 3r,4r,4ar,6as,8r,9r,11as,11bs-4,9-bis hydroxymethyl-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta a naphthalene-3,9-diol, 8,11a-methano-11ah-cyclohepta a naphthalene-4,9-dimethanol, tetradecahydro-3,9-dihydroxy-4,11b-dimethyl-, 3r-3-alpha,4-alpha,4a-alpha,6a-beta,8-beta,9-beta,11a-beta,11b-beta, 9,15-cyclo-c,18-dinor-14,15-secoandrostane-4,17-dimethanol, 3,17-dihydroxy-4-methyl-, 3alpha,4alpha,5alpha,17alpha, aphidicolin, aphidicolin, +/-, ccris 1783, chembl29711, unii-192tj6pp19 |
PubChem CID | 457964 |
ChEBI | CHEBI:2766 |
SMILES | CC12CCC(C(C1CCC3C24CCC(C(C3)C4)(CO)O)(C)CO)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC611970050
|
Thermo Fisher Scientific
611970050 |
5mg |
Each for $671.56
|
|
Chemical Identifiers
38966-21-1 | |
338.488 | |
NOFOAYPPHIUXJR-APNQCZIXSA-N | |
457964 | |
CC12CCC(C(C1CCC3C24CCC(C(C3)C4)(CO)O)(C)CO)O |
C20H34O4 | |
MFCD00083214 | |
+-aphidicolin, +/--aphidicolin, 3r,4r,4ar,6as,8r,9r,11as,11bs-4,9-bis hydroxymethyl-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta a naphthalene-3,9-diol, 8,11a-methano-11ah-cyclohepta a naphthalene-4,9-dimethanol, tetradecahydro-3,9-dihydroxy-4,11b-dimethyl-, 3r-3-alpha,4-alpha,4a-alpha,6a-beta,8-beta,9-beta,11a-beta,11b-beta, 9,15-cyclo-c,18-dinor-14,15-secoandrostane-4,17-dimethanol, 3,17-dihydroxy-4-methyl-, 3alpha,4alpha,5alpha,17alpha, aphidicolin, aphidicolin, +/-, ccris 1783, chembl29711, unii-192tj6pp19 | |
CHEBI:2766 |
Specifications
Pass Test | |
5mg | |
C20H34O4 | |
+-aphidicolin, +/--aphidicolin, 3r,4r,4ar,6as,8r,9r,11as,11bs-4,9-bis hydroxymethyl-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta a naphthalene-3,9-diol, 8,11a-methano-11ah-cyclohepta a naphthalene-4,9-dimethanol, tetradecahydro-3,9-dihydroxy-4,11b-dimethyl-, 3r-3-alpha,4-alpha,4a-alpha,6a-beta,8-beta,9-beta,11a-beta,11b-beta, 9,15-cyclo-c,18-dinor-14,15-secoandrostane-4,17-dimethanol, 3,17-dihydroxy-4-methyl-, 3alpha,4alpha,5alpha,17alpha, aphidicolin, aphidicolin, +/-, ccris 1783, chembl29711, unii-192tj6pp19 | |
CC12CCC(C(C1CCC3C24CCC(C(C3)C4)(CO)O)(C)CO)O | |
457964 | |
338.49 | |
Aphidicolin |
226°C to 235°C | |
38966-21-1 | |
MFCD00083214 | |
NOFOAYPPHIUXJR-APNQCZIXSA-N | |
338.488 | |
CHEBI:2766 | |
98% min. |
Safety and Handling
RTECSNumber : PB9185000