Please login to your online account to display your discounted pricing

Alfa Aesar™ Apelin-36, Human

For cellular and molecular biology applications

Specifications

Product Type Apelin-36, human
Cross Reactivity Human
Molecular Weight 4195.82
Storage Requirements -30° to -10°C
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66238MCR

 
alfa aesar™
J66238MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66238LB0

 
alfa aesar™
J66238LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Apelin-36, human
Cross Reactivity Human
Molecular Weight 4195.82
Storage Requirements -30° to -10°C
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC