Please login to your online account to display your discounted pricing

Alfa Aesar™ Apelin-13, Human, Bovine

For cellular and molecular biology applications

Specifications

Product Type Apelin-13, human, bovine
Cross Reactivity Human, Bovine
Molecular Weight 1550.82
Storage Requirements -30° to -10°C
Sequence Gln-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66620EXD

 
alfa aesar™
J66620EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66620MCR

 
alfa aesar™
J66620MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Apelin-13, human, bovine
Cross Reactivity Human, Bovine
Molecular Weight 1550.82
Storage Requirements -30° to -10°C
Sequence Gln-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC