Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ APC2 (Anaphase-Promoting Complex) Ab-1, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ APC2 (Anaphase-Promoting Complex) Ab-1, Rabbit Polyclonal Antibody.

$554.55 - $1,716.27

Specifications

Antigen APC2 (Anaphase-Promoting Complex) Ab-1
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB067P

 
thermo scientific™ lab vision™
RB067P
Purified Ab with BSA and Azide 1mL Each for $1,561.82

RB067P0

 
thermo scientific
RB067P0
Purified Ab with BSA and Azide 100µL Each for $554.55

RB067P1

 
thermo scientific
RB067P1
Purified Ab with BSA and Azide 500µL Each for $1,143.57

RB067P1ABX

 
thermo scientific™ lab vision™
RB067P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $1,192.24

RB067PABX

 
thermo scientific™ lab vision™
RB067PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,716.27
Description & Specifications

Specifications

Antigen APC2 (Anaphase-Promoting Complex) Ab-1
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Cell Cycle
Type Primary
Immunogen A peptide from C-terminus of the human APC2
Isotype IgG

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ APC2 (Anaphase-Promoting Complex) Ab-1, Rabbit Polyclonal Antibody.

APC2 is a cullin-related subunit of the anaphase-promoting complex (APC), or cyclosome. APC2 specifically interacts with APC11, another APC subunit.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: aa 810-822

Immunogen: A peptide from C-terminus of the human APC2

Molecular Weight: 92kDa

Positive Control: HeLa cells

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO