Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ APC2 (Anaphase-Promoting Complex) Ab-1, Rabbit Polyclonal Antibody


Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ APC2 (Anaphase-Promoting Complex) Ab-1, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB067P1ABX

 View more versions of this product

Catalog No. RB067P1ABX

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Regulatory Status RUO
Applications Western Blotting
Cross Reactivity Human
Antigen APC2 (Anaphase-Promoting Complex) Ab-1
Immunogen A peptide from C-terminus of the human APC2
Format Concentrated
Host Species Rabbit
Quantity 500µL
Classification Polyclonal
Concentration 1mg/mL
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Conjugate Unlabeled
Research Discipline Cell Cycle

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ APC2 (Anaphase-Promoting Complex) Ab-1, Rabbit Polyclonal Antibody.

APC2 is a cullin-related subunit of the anaphase-promoting complex (APC), or cyclosome. APC2 specifically interacts with APC11, another APC subunit.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not tested.

Epitope: aa 810-822

Immunogen: A peptide from C-terminus of the human APC2

Molecular Weight: 92kDa

Positive Control: HeLa cells

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO