Please login to your online account to display your discounted pricing

Thermo Scientific™ ApaI

Cut at GGGCC^C sites with ApaI restriction enzyme, which functions best at 37°C in B buffer (Isoschizomers: PspOMI).

$89.75 - $89.75

Specifications

Components 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2 and 0.1mg/mL BSA
Concentration 10U/µL
Incubator Temperature 37°C
Product Name Apal
View More Specs

 Disclaimers

Patent pending.

The purchase of this product includes a non-transferable license to use this product for the purchaser's internal research. All other commercial uses of this product, including without limitation product use for diagnostic purposes, resale of product in the original or any modified form or product use in providing commercial services require a separate license from Fermentas.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

FERER1415

 
thermo scientific™
ER1415
5,000U Each for $89.75
Description & Specifications

Specifications

Components 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2 and 0.1mg/mL BSA
Concentration 10U/µL
Incubator Temperature 37°C
Product Name Apal

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities