Please login to your online account to display your discounted pricing

Invitrogen™ Anza™ T4 DNA Ligase Master Mix DFS Item


Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Manufacturer: invitrogen™  LSIVGN2108

 View more versions of this product

Anza™ T4 DNA Ligase Master Mix

 Disclaimers

For research use only. Not for use in diagnostic procedures.

Catalog No. IVGN2108

  • / Each
 

Description & Specifications

Specifications

Quantity No. of Reactions: 200
Includes Anza™ T4 DNA Ligase Master Mix
Storage Requirements Store at -20°C
For Use With (Application) Cloning, Gene Expression/RNA Analysis, Protein Expression, Genotyping and Genetic Variation, RNAi, Sequencing
Concentration 4x

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One Ligase for both cohesive and blunt end ligations
• 4X conc. enables higher volume of dilute DNA in reaction mixture
• Ligation complete in 15 minutes
• Convenient Master Mix format reduces pipetting

Certifications

CoA