Invitrogen™ Anza™ T4 DNA Ligase Master Mix DFS Item
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Supplier:  Invitrogen™ IVGN2104
 View more versions of this product
Catalog No. IVGN2104
Includes: Anza™ T4 DNA Ligase Master Mix
Description
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One Ligase for both cohesive and blunt end ligations
• 4X conc. enables higher volume of dilute DNA in reaction mixture
• Ligation complete in 15 minutes
• Convenient Master Mix format reduces pipetting
Certifications
CoA
Specifications
Specifications
4X
Approved for shipment on Wet or Dry Ice
Elution Buffer, 1X Anza Buffer
T4 DNA Ligase Master Mix
250 μL Anza T4 DNA Ligase Master Mix
Store at -5 to -30°C.
T4 DNA Ligase
50 Reactions
Anza™
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.