missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Anza™ Alkaline Phosphatase DFS Item

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Manufacturer:  Invitrogen™ LSIVGN2208

 View more versions of this product

Catalog No. IVGN2208


 

Disclaimers:

For research use only. Not for use in diagnostic procedures.


Includes: Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer

Specifications

Description

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• Utilizes the Anza 10X Buffer
• One-step digestion and dephosphorylation protocol
• Two-step dephosphorylated protocols available
• Heat inactivated in 5 minutes

Certifications

CoA

Description & Specifications

Specifications

1U/μL
No. of Reactions: 2000
Store at -20°C
Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Cloning, Gene Expression/RNA Analysis, Protein Expression, Genotyping and Genetic Variation, RNAi, Sequencing
Promotions

Promotions