Please login to your online account to display your discounted pricing

Invitrogen™ Anza™ Alkaline Phosphatase DFS Item


Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Manufacturer: invitrogen™  LSIVGN2204

 View more versions of this product

Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer

 Disclaimers

For research use only. Not for use in diagnostic procedures.

Catalog No. IVGN2204

  • / Each
 

Description & Specifications

Specifications

Includes Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Storage Requirements Store at -20°C
Concentration 1U/μL
Quantity No. of Reactions: 500
For Use With (Application) Cloning, Gene Expression/RNA Analysis, Protein Expression, Genotyping and Genetic Variation, RNAi, Sequencing

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• Utilizes the Anza 10X Buffer
• One-step digestion and dephosphorylation protocol
• Two-step dephosphorylated protocols available
• Heat inactivated in 5 minutes

Certifications

CoA