Please login to your online account to display your discounted pricing

Invitrogen™ Anza™ 26 Eco32I DFS Item


Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Manufacturer: invitrogen™  LSIVGN0268

 View more versions of this product

Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer

 Disclaimers

For research use only. Not for use in diagnostic procedures.

Catalog No. IVGN0268

  • / Each
 

Description & Specifications

Specifications

Includes Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Quantity 750μL
Units 15000 Units
For Use With (Application) Cloning, Gene Expression/RNA Analysis, Protein Expression, Genotyping and Genetic Variation, RNAi, Sequencing
Storage Requirements Store at -20°C
Cut Site GAT^ATC, GATATC
Concentration 20U/μL

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity

Certifications

CoA