missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Anza™ 22 SmaI DFS Item

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

$87.00 - $318.00

Specifications

Concentration 5U/μL
Cut Site CCC^GGG, CCCGGG
For Use With (Application) Restriction enzyme cloning
Storage Requirements –20°C
View More Specs

Disclaimers:

For research use only. Not for use in diagnostic procedures.


Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
IVGN0226 Invitrogen™
LSIVGN0226
400μL Each for $87.00
IVGN0228 Invitrogen™
LSIVGN0228
8000 Unit(s) Each for $318.00
Specifications

Description

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity

Certifications

CoA

Description & Specifications

Specifications

5U/μL
CCC^GGG, CCCGGG
Restriction enzyme cloning
–20°C