Please login to your online account to display your discounted pricing

Invitrogen™ Anza™ 22 SmaI DFS Item


Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Manufacturer: invitrogen™  LSIVGN0228

 View more versions of this product

Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer

 Disclaimers

For research use only. Not for use in diagnostic procedures.

Catalog No. IVGN0228

  • / Each
 

Description & Specifications

Specifications

Units 8000 Unit(s)
For Use With (Application) Restriction enzyme cloning
Storage Requirements –20°C
Storage Buffer Anza™ 10x buffer, Anza™ 10x red buffer
Product Name Anza™ 22 SmaI Enzyme
Quantity 8000 Unit(s)
Product Type Anza™ 22 SmaI Enzyme
Includes 1.6mL Anza 22 SmaI, 2mL Anza buffer (10X), 2mL Anza red buffer (10X)
Cut Site CCC^GGG, CCCGGG
Concentration 5U/μL

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity

Certifications

CoA