missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Anza™ 20 SacI DFS Item

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

$79.50 - $290.00

Specifications

Concentration 10U/μL
Cut Site GAGCT^C, GAGCTC
For Use With (Application) Cloning, Gene Expression/RNA Analysis, Protein Expression, Genotyping and Genetic Variation, RNAi, Sequencing
Storage Requirements Store at -20°C
View More Specs

Disclaimers:

For research use only. Not for use in diagnostic procedures.


Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
IVGN0206 Invitrogen™
LSIVGN0206
160μL Each for $79.50
IVGN0208 Invitrogen™
LSIVGN0208
800μL Each for $290.00
Specifications

Description

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity

Certifications

CoA

Description & Specifications

Specifications

10U/μL
GAGCT^C, GAGCTC
Cloning, Gene Expression/RNA Analysis, Protein Expression, Genotyping and Genetic Variation, RNAi, Sequencing
Store at -20°C