Please login to your online account to display your discounted pricing

Invitrogen™ Anza™ 1 NotI DFS Item


Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Manufacturer: invitrogen™  LSIVGN0016

 View more versions of this product

Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer

 Disclaimers

For research use only. Not for use in diagnostic procedures.

Catalog No. IVGN0016

  • / Each
 

Description & Specifications

Specifications

Quantity 2000 Unit(s)
Storage Requirements –20°C
Includes 100μL Anza 1 NotI, 500μL Anza buffer (10X), 500μL Anza red buffer (10X)
For Use With (Application) Restriction enzyme cloning
Units 2000 Unit(s)
Product Type Anza™ 1 NotI Enzyme
Product Name Anza™ 1 NotI Enzyme
Storage Buffer Anza™ 10x buffer, Anza™ 10x red buffer
Concentration 20U/μL
Cut Site GC^GGCCGC, GCGGCCGC

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity

Certifications

CoA