Please login to your online account to display your discounted pricing

Alfa Aesar™ Antisauvagine-30

For cellular and molecular biology applications

$534.14 - $691.14

Specifications

Product Type Antisauvagine-30
Molecular Weight 3650.25
Storage Requirements -30° to -10°C
Sequence DPhe-His-Leu-Leu-Arg-Lys-Met-Ile-Glu-Ile-Glu-Lys-Gln-Glu-Lys-Glu-Lys-Gln-Gln-Ala-Ala-Asn-Asn-Arg-Leu-Leu-Leu-Asp-Thr-Ile
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66055EXD

 
alfa aesar™
J66055EXD
0.5mg Each for $534.14

AAJ66055MCR

 
alfa aesar™
J66055MCR
1mg Each for $691.14
Description & Specifications

Specifications

Product Type Antisauvagine-30
Molecular Weight 3650.25
Storage Requirements -30° to -10°C
Sequence DPhe-His-Leu-Leu-Arg-Lys-Met-Ile-Glu-Ile-Glu-Lys-Gln-Glu-Lys-Glu-Lys-Gln-Gln-Ala-Ala-Asn-Asn-Arg-Leu-Leu-Leu-Asp-Thr-Ile
Format Solid
Regulatory Status RUO
Purification Method HPLC