Please login to your online account to display your discounted pricing

Antipyrine, 99%, ACROS Organics™


Quantity 5kg

Manufacturer: thermo fisher scientific  AC104970050

 View more versions of this product

Catalog No. AC104970050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 188.23
Quantity 5kg
Color White
EINECS Number 200-486-6
TSCA TSCA
Melting Point 109° to 113°C
RTECS Number CD2450000
Chemical Name or Material Antipyrine
Name Note 99%
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C11H12N2O
Packaging Plastic drum
MDL Number MFCD00003146
CAS 60-80-0
Synonym 2, 3-Dimethyl-1-phenyl-3-pyrazolin-5-one
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Antipyrine
Name Note 99%
CAS 60-80-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H12N2O
Formula Weight 188.23
MDL Number MFCD00003146
Synonym 2, 3-Dimethyl-1-phenyl-3-pyrazolin-5-one
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents