Please login to your online account to display your discounted pricing

Alfa Aesar™ Antipain dihydrochloride

Chemical Identifiers

Chemical Name or Material Antipain dihydrochloride
CAS 37682-72-7
Formula Weight 677.6
MDL Number MFCD00135957
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63680LB0

  SDS
alfa aesar™
J63680LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 677.6
Chemical Name or Material Antipain dihydrochloride
CAS 37682-72-7
Physical Form Powder
MDL Number MFCD00135957
Color White to pale yellow
Quantity 5mg
Concentration 0.01M in water
Storage Note 1 -30° to -10°C
RTECS Number YV9350800

Applications
Antipain inhibits trypsin, papain, and catherpsins A and B (a reversible inhibitor of cysteine and serine proteases). It is used to evaluate the role of proteases in cell transformations. It is used to help identify new proteases.

Solubility
0.01M in water

Notes
Store at -20°C . Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

-30° to -10°C