Please login to your online account to display your discounted pricing

Antimony(V) oxide, White lump, 99.99% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Antimony(V) oxide
CAS 1309-64-4
Physical Form Lump
Assay Percent Range 99.99% (Metals basis)
Molecular Formula Sb2O3
Formula Weight 291.5
MDL Number MFCD00011214
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3631418

  SDS
alfa aesar
3631418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3631430

  SDS
alfa aesar
3631430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lump
Formula Weight 291.5
Quantity 50g
Color White
Chemical Name or Material Antimony(V) oxide
EINECS Number 215-175-0
Odor Odorless
Assay Percent Range 99.99% (Metals basis)
Molecular Formula Sb2O3
MDL Number MFCD00011214
CAS 1309-64-4
Health Hazard 1 H351: Suspected of causing cancer.
Health Hazard 2 Do not breathe dust. Wear suitable protective clothing and gloves
Density 5.67g/cm3
Safety and Handling

H351: Suspected of causing cancer.

Do not breathe dust. Wear suitable protective clothing and gloves