missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony(V) oxide, 99.998% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1064306

 View more versions of this product

Catalog No. AA1064306



Specifications

Specifications

Antimony(V) oxide
99.99% (Metal basis)
MFCD00011216
antimonic oxide
O=[Sb](=O)O[Sb](=O)=O
323.515
323.5
Odorless
5g
1314-60-9
O5Sb2
Yellowish powder
LJCFOYOSGPHIOO-UHFFFAOYSA-N
(dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane
14813
Powder
Yellow

Chemical Identifiers

1314-60-9
323.515
LJCFOYOSGPHIOO-UHFFFAOYSA-N
14813
O=[Sb](=O)O[Sb](=O)=O
O5Sb2
MFCD00011216
antimonic oxide
(dioxo-$l^{5}-stibanyl)oxy-dioxo-$l^{5}-stibane
Safety and Handling

Safety and Handling

H302-H332-H411

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 215-237-7

Documents