Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(V) oxide, 30% in H2O, colloidal dispersion

Flame retardant

Chemical Identifiers

Chemical Name or Material Antimony(V) oxide
CAS 1314-60-9
Name Note Colloidal dispersion
Formula Weight 323.5
MDL Number MFCD00011216
Synonym Antimony pentoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4012836

  SDS
alfa aesar™
4012836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40128A4

  SDS
alfa aesar™
40128A4
2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Particles In Liquid
Formula Weight 323.5
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Antimony(V) oxide
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
TSCA TSCA
Particle Size 0.03 Micron
Health Hazard 1 Danger
MDL Number MFCD00011216
Synonym Antimony pentoxide
Quantity 500g
Odor Odorless
Name Note Colloidal dispersion
Concentration 30% in H2O
CAS 1314-60-9
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.