Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(V) chloride, ultra dry, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Antimony(V) chloride
CAS 7647-18-9
Name Note Ultra dry
Formula Weight 299.02
MDL Number MFCD00011213
UN Number UN1730
Merck Index 14,695
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3567703

  SDS
alfa aesar™
3567703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3567709

  SDS
alfa aesar™
3567709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid, Ampouled Under Argon
Formula Weight 299.02
MDL Number MFCD00011213
Density 2.336g/mL
Refractive Index 1.601
Assay Percent Notes (metals basis)
RTECS Number CC5075000
Odor Pungent
Sensitivity Moisture Sensitive
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Melting Point 2.8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANTIMONY PENTACHLORIDE, LIQUID
Boiling Point 79°C (22mmHg)
Chemical Name or Material Antimony(V) chloride
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99.999%
UN Number UN1730
Merck Index 14,695
CAS 7647-18-9
Health Hazard 1 Danger
TSCA TSCA
Quantity 1g
Name Note Ultra dry
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.