Learn More
Antimony(V)-chloride, 99%, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 201212500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
92.0°C (30.0 mmHg) | |
2.8°C | |
7647-18-9 | |
Liquid | |
98.5% min | |
SbCl5 | |
01,42; 04,21; 05,18; 06,22; 07,12; 15,14; 17,17 | |
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german | |
QFJJIKGZKLCYFT-UHFFFAOYSA-N | |
pentachloro-$l^{5}-stibane | |
24294 | |
99% | |
2.36 |
Antimony(V)-chloride | |
99% | |
Colorless to Yellow | |
250 g | |
Cl5H8Sb | |
MFCD00011213 | |
15, 686 | |
Solubility in water: reacts with water forming hcl and antimonyoxide. Other solubilities: soluble in hcl, carbon disulfide, dichloromethane, soluble in an aqueous solution of tartaric acid,, ccl4 and chloroform | |
Cl.Cl.Cl.Cl.Cl.[SbH3] | |
307.07 | |
299.01 | |
Glass bottle | |
2.3600g/mL |
Chemical Identifiers
7647-18-9 | |
307.07 | |
QFJJIKGZKLCYFT-UHFFFAOYSA-N | |
24294 | |
Cl.Cl.Cl.Cl.Cl.[SbH3] |
Cl5H8Sb | |
MFCD00011213 | |
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german | |
pentachloro-$l^{5}-stibane |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 231-601-8
RTECSNumber : CC5075000
TSCA : TSCA
Recommended Storage : Product may develop some turbidity or precipitate
RUO – Research Use Only