Please login to your online account to display your discounted pricing

Antimony(V)-chloride, 99%, anhydrous, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  201210500

 View more versions of this product

Catalog No. AC201210500

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 299.01
Assay ≥98.5% (SbCl5)
Physical Form Liquid
Quantity 50g
Color Colorless to yellow
Merck Index 15, 686
TSCA TSCA
EINECS Number 231-601-8
RTECS Number CC5075000
Density 2.3600g/mL
Fieser 01,42; 04,21; 05,18; 06,22; 07,12; 15,14; 17,17
Molecular Formula Cl5Sb
CAS 7647-18-9
Health Hazard 1 Danger
Chemical Name or Material Antimony(V)-chloride
Packaging Glass bottle
Storage Information Product may develop some turbidity or precipitate
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment.
Health Hazard 2 GHS P Statement Toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. May cause respiratory irritation.
Assay Percent Range 99%
Name Note 99%
Linear Formula SbCl5
MDL Number MFCD00011213

Specifications

Chemical Name or Material Antimony(V)-chloride
Name Note 99%
CAS 7647-18-9
Assay ≥98.5% (SbCl5)
Assay Percent Range 99%
Molecular Formula Cl5Sb
Linear Formula SbCl5
Formula Weight 299.01
MDL Number MFCD00011213
Accessories

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

GHS P Statement Toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. May cause respiratory irritation.

SDS & Safety Documents