Please login to your online account to display your discounted pricing

Antimony(V) chloride, 99%, Alfa Aesar™

$79.55 - $147.66

Chemical Identifiers

Chemical Name or Material Antimony(V) chloride
CAS 7647-18-9
Physical Form Liquid
Assay Percent Range 99%
Molecular Formula SbCl5
Formula Weight 299.02
MDL Number MFCD00011213
Synonym Antimony pentachloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4197818

  SDS
alfa aesar
4197818
50g Each for $79.55

AA4197830

  SDS
alfa aesar
4197830
250g Each for $147.66

AA41978A1

  SDS
alfa aesar
41978A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 299.02
Quantity 50g
Color Red-yellow
DOT Information DOT Class: 8, Packaging Group: II
Density 2.336g/cm3
EINECS Number 231-601-8
Appearance Red-yellow liquid
Assay Percent Range 99%
Synonym Antimony pentachloride
Chemical Name or Material Antimony(V) chloride
Odor Pungent
Molecular Formula SbCl5
Physical Form Liquid
MDL Number MFCD00011213
CAS 7647-18-9
Health Hazard 1 H314-H411
Safety and Handling

H314-H411