missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(V) chloride, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 1757014

 View more versions of this product

Catalog No. AA1757014



Specifications

Specifications

Antimony(V) chloride
(metals basis)
SbCl5
UN1730
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
Cl[Sb](Cl)(Cl)(Cl)Cl
299.01
299.02
99.997%
Pungent
Moisture Sensitive
Red-Yellow
25g
7647-18-9
Cl5Sb
MFCD00011213
14,695
VMPVEPPRYRXYNP-UHFFFAOYSA-I
pentachloro-$l^{5}-stibane
24294
Liquid
2.336g/mL
1.601
79°C (22mmHg)
2.8°C

Chemical Identifiers

7647-18-9
299.01
VMPVEPPRYRXYNP-UHFFFAOYSA-I
24294
Cl[Sb](Cl)(Cl)(Cl)Cl
Cl5Sb
MFCD00011213
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
pentachloro-$l^{5}-stibane
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANTIMONY PENTACHLORIDE, LIQUID

RTECSNumber : CC5075000

TSCA : TSCA

SDS Documents