Learn More
Antimony(V) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01757022
Description
Antimony(V) chloride is used as a catalyst and an analytical reagent for testing alkaloids and cesium. It is also used as a polymerization catalyst as well as involved in the chlorination of organic compounds. It acts as a Lewis acid and a strong oxidizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
79°C (22mmHg) | |
2.8°C | |
Red-Yellow | |
100 g | |
Cl5H8Sb | |
MFCD00011213 | |
14,695 | |
Miscible with chloroform,hydrochloric acid,tartaric acid,carbon disulfide and carbon tetrachloride. | |
Cl.Cl.Cl.Cl.Cl.[SbH3] | |
307.07 | |
299.02 | |
Pungent | |
Moisture sensitive |
Antimony(V) chloride | |
7647-18-9 | |
Liquid | |
(metals basis) | |
SbCl5 | |
UN1730 | |
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german | |
QFJJIKGZKLCYFT-UHFFFAOYSA-N | |
pentachloro-$l^{5}-stibane | |
24294 | |
99.997% | |
1.601 | |
2.336 g/mL |
Chemical Identifiers
7647-18-9 | |
307.07 | |
QFJJIKGZKLCYFT-UHFFFAOYSA-N | |
24294 | |
Cl.Cl.Cl.Cl.Cl.[SbH3] |
Cl5H8Sb | |
MFCD00011213 | |
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german | |
pentachloro-$l^{5}-stibane |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P270-P271-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANTIMONY PENTACHLORIDE, LIQUID
EINECSNumber : 231-601-8
RTECSNumber : CC5075000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only