Please login to your online account to display your discounted pricing

Antimony Trioxide (Powder/Certified), Fisher Chemical

$431.77 - $431.77

Chemical Identifiers

Chemical Name or Material Antimony Trioxide
CAS 1309-64-4
Purity Grade Certified Grade
Physical Form Powder/Solid
Molecular Formula O3Sb2
Formula Weight 291.52
MDL Number MFCD00011214
Synonym Antimonous Oxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A860-500

  SDS
fisher scientific company
FLA860500
Glass Bottle 500g Each for $431.77
Case of 6 Each for $2,590.62
Description & Specifications

Specifications

Formula Weight 291.52
Boiling Point 1550°C
Melting Point 656°C
Purity Grade Certified Grade
Physical Form Powder/Solid
Quantity 500g
Molecular Formula O3Sb2
Color White
Synonym Antimonous Oxide
Lead (Pb) 0.04% max.
Arsenic (As) 0.20% max.
Chemical Name or Material Antimony Trioxide
CAS 1309-64-4
Chloride 0.005% max.
MDL Number MFCD00011214
Identification Pass Test
Sulfate 0.01% max.
Iron (Fe) 0.01% max.
Packaging Glass Bottle
ChemAlert Storage Symbol Gray
Safety and Handling