Learn More
Antimony trichloride, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 401371000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
223.0°C | |
73.0°C | |
White | |
100 g | |
SbCl3 | |
08,19 | |
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
trichlorostibane | |
24814 | |
228.11 | |
ACS Reagent |
Antimony trichloride | |
10025-91-9 | |
Powder, Crystals and/or Chunks | |
Cl3Sb | |
MFCD00011212 | |
15,696 | |
Solubility in water: 100g/L (25°C). Other solubilities: soluble in diluted hcl,, soluble in chloroform (+/- 22%), soluble in alcohol, benzene, carbon disulfide,, acetone, tartaric acid, ccl4, insoluble in pyridine, quinoline, organic bases | |
[Cl-].[Cl-].[Cl-].[Sb+3] | |
228.11 | |
CHEBI:74856 | |
≥99.0% (Iodimetry) | |
Glass bottle |
Chemical Identifiers
10025-91-9 | |
228.11 | |
FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
24814 | |
[Cl-].[Cl-].[Cl-].[Sb+3] |
Cl3Sb | |
MFCD00011212 | |
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
CHEBI:74856 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 233-047-2
RTECSNumber : CC4900000
TSCA : TSCA
RUO – Research Use Only