missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony trichloride, 99.5%, ACROS Organics™

Manufacturer:  Acros Organics AC201680100

 View more versions of this product

Catalog No. AC201680100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Antimony trichloride
99.5%
SbCl3
08,19
antimonous chloride
Solubility in water: 100g/L (25°C). Other solubilities: soluble in diluted hcl,,soluble in chloroform (+/- 22%),soluble in alcohol,benzene,carbon disulfide,,acetone,tartaric acid,ccl4,insoluble in pyridine,quinoline,organic bases
Cl[Sb](Cl)Cl
228.11
CHEBI:74856
Liquid
0.01% max. (in HCl)
Plastic Bottle
Colorless
10kg
10025-91-9
Cl3Sb
MFCD00011212
15, 696
100ppm max.
FAPDDOBMIUGHIN-UHFFFAOYSA-K
trichlorostibane
24814
228.11
≥99.4% (Iodimetry)
10ppm max.
223°C
73°C

Chemical Identifiers

10025-91-9
228.11
FAPDDOBMIUGHIN-UHFFFAOYSA-K
24814
trichlorostibane
Cl3Sb
MFCD00011212
antimonous chloride
CHEBI:74856
Cl[Sb](Cl)Cl
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents