Please login to your online account to display your discounted pricing

Antimony Standard (Trace ICP/ICP-MS Grade), SPEX CertiPrep™

$124.11 - $124.11

Chemical Identifiers

Chemical Name or Material Antimony Standard
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

CLSB72M

 
spex certiprep llc
SPXCLSB72M
30mL Each for $124.11
Description & Specifications

Specifications

Quantity 30mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Chemical Name or Material Antimony Standard
Health Hazard 1 Causes skin irritation, serious eye irritation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Solvent or Matrix H2O/0.6% Tart. Acid/Tr. HNO3
DOT Information Nonhazardous
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Causes skin irritation, serious eye irritation

Ambient