missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony Standard For ICP, 10,000μg/mL in H2O/6% Tart. Acid/1% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$286.69 - $498.40

Chemical Identifiers

For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Chemical Name or Material Antimony Standard
View More Specs

Disclaimers:

Product intended for laboratory use only


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
PLSB7-3X Spex Certiprep Llc
PLSB73X
Triple-leached LDPE bottle 500mL Each for $498.40
PLSB7-3Y Spex Certiprep Llc
PLSB73Y
Triple-leached LDPE bottle 125mL Each for $286.69
Specifications

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sb10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Antimony Standard

Specifications

87-69-4,7697-37-2,7440-36-0,7732-18-5
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
H2O/6% Tart. Acid/1% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Assurance Grade
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
500mL
Antimony Standard

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient