missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony rod, 12.7mm (0.500 in.) dia., 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4718113

 View more versions of this product

Catalog No. AA4718113



Specifications

Specifications

Antimony rod
20g
7440-36-0
(metals basis)
MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
5354495
Cast Rod
≈2.16cm/20g
12.7mm (0.500 in.) dia.
Odorless
Sb
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
[Sb]
121.76
CHEBI:30304
99.999%

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
antimony
Sb
MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
CHEBI:30304
[Sb]
Safety and Handling

Safety and Handling

RTECSNumber : CC4025000

TSCA : TSCA

SDS Documents