Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony rod, 12.7mm (0.500 in.) dia., 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Antimony rod
CAS 7440-36-0
Name Note 12.7mm (0.500 in.) dia.
MDL Number MFCD00134030
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4718113

  SDS
alfa aesar™
4718113
20g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4718120

  SDS
alfa aesar™
4718120
80g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 20g
Assay Percent Notes (metals basis)
MDL Number MFCD00134030
Chemical Name or Material Antimony rod
Name Note 12.7mm (0.500 in.) dia.
CAS 7440-36-0
Assay 99.999%
Weight ≈2.16cm/20g
Odor Odorless
TSCA TSCA
Physical Form Cast Rod
RTECS Number CC4025000