missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony rod, 12.7mm (0.50 in.) dia. x 10cm (3.9 in.) long, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 10098KG

 View more versions of this product

Catalog No. AA10098KG



Specifications

Specifications

Antimony rod
1 pc.
7440-36-0
(metals basis)
MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
5354495
Cast Rod
≈81.2g (0.178lbs)/10cm piece
12.7mm (0.50 in.) dia. x 10cm (3.9 in.) long
Odorless
Sb
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
[Sb]
121.76
CHEBI:30304
99.8%

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
antimony
Sb
MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
CHEBI:30304
[Sb]
Safety and Handling

Safety and Handling

RTECSNumber : CC4025000

TSCA : TSCA

Documents