Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony rod, 10-12mm (0.39-0.47 in.) dia., 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Antimony rod
CAS 7440-36-0
Name Note 10 to 12mm (0.39 to 0.47 in.) dia.
MDL Number MFCD00134030
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1106913

 
alfa aesar™
11069-13
20g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Weight ≈2.7cm/20g
Quantity 20g
Chemical Name or Material Antimony rod
Name Note 10 to 12mm (0.39 to 0.47 in.) dia.
CAS 7440-36-0
Physical Form Cast rod
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00134030
RTECS Number CC4025000
TSCA TSCA