Please login to your online account to display your discounted pricing

Antimony, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SA450500

 View more versions of this product

Catalog No. SA450-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Color Colorless
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
Viscosity [NULL]
Density 1.0g/cm³
pH 1
Traceability to NIST Lot Number Used
CAS 7732-18-5
ChemAlert Storage Symbol White
Chemical Name or Material Antimony, Reference Standard Solution
Concentration 990 to 1010ppm
MDL Number MFCD00134030
CAS 10025-91-9
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Packaging Poly Bottle
Name Note 1000ppm ±1%
CAS 7647-01-0

Specifications

Chemical Name or Material Antimony, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 10025-91-9
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134030
Accessories
 

SDS & Safety Documents