missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Antimony powder, -200 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
$58.96 - $579.51
Chemical Identifiers
CAS | 7440-36-0 |
---|---|
Molecular Formula | Sb |
Molecular Weight (g/mol) | 121.76 |
MDL Number | MFCD00011211 MFCD00134030 |
InChI Key | WATWJIUSRGPENY-UHFFFAOYSA-N |
Synonym | powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german |
PubChem CID | 5354495 |
ChEBI | CHEBI:30304 |
IUPAC Name | antimony |
SMILES | [Sb] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-36-0 | |
121.76 | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
5354495 | |
antimony |
Sb | |
MFCD00011211 MFCD00134030 | |
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german | |
CHEBI:30304 | |
[Sb] |
Specifications
Gray | |
Gray powder | |
Sb | |
1 g | |
WATWJIUSRGPENY-UHFFFAOYSA-N | |
antimony | |
5354495 | |
-200 Mesh | |
Powder |
7440-36-0 | |
99.999% (Metals basis) | |
MFCD00011211 MFCD00134030 | |
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german | |
[Sb] | |
121.76 | |
CHEBI:30304 | |
Odorless | |
Antimony |
Safety and Handling
P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c
H335-H351-H373
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 231-146-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only