missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony powder, -200 mesh, 99.999% (metal basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1464006

 View more versions of this product

Catalog No. AA1464006



Specifications

Specifications

Antimony
99.999% (Metals basis)
MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
[Sb]
121.76
CHEBI:30304
6.68g/cm3
Gray
7440-36-0
Sb
Gray powder
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
5354495
Powder
Odorless
5g

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
antimony
Sb
MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
CHEBI:30304
[Sb]
Safety and Handling

Safety and Handling

H301-H331-H411-H401: Toxic if swallowed. Toxic if inhaled. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 231-146-5

SDS Documents