missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01009936

 View more versions of this product

Catalog No. AA1009936



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
7440-36-0
Sb
UN2871
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
[Sb]
121.76
CHEBI:30304
−200 Mesh
Powder
Antimony powder
(metals basis)
MFCD00011211 MFCD00134030
500 g
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
5354495
99.5%
Odorless

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
[Sb]
Sb
MFCD00011211 MFCD00134030
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
CHEBI:30304
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c

H335-H351-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY POWDER

EINECSNumber : 231-146-5

RTECSNumber : CC4025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only