missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony powder, -100 mesh, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 10100A1

 View more versions of this product

Catalog No. AA10100A1



Specifications

Specifications

Antimony powder
Apparent density: 2.4g/cm3 to 2.9g/cm3
1kg
Odorless
Sb
UN2871
WATWJIUSRGPENY-UHFFFAOYSA-N
antimony
5354495
Powder
−100 Mesh
1380°C
7440-36-0
(metals basis)
MFCD00134030
0vkz97k3ub
[Sb]
121.76
CHEBI:30304
99.5%

Chemical Identifiers

7440-36-0
121.76
WATWJIUSRGPENY-UHFFFAOYSA-N
5354495
antimony
Sb
MFCD00134030
0vkz97k3ub
CHEBI:30304
[Sb]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P301+P310-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY POWDER

TSCA : TSCA

Documents