missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Antimony Potassium Tartrate Trihydrate (USP), Fisher Chemical™
$349.67 - $627.27

Chemical Identifiers
CAS | 28300-74-5 |
---|---|
Molecular Formula | C8H4K2O12Sb2 · 3 H2O |
MDL Number | MFCD00167056 |
Synonym | Tartrated Antimony |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A867-250
|
Fisher Chemical
FLA867250 |
250g | Glass Bottle |
Each for $349.67
Case of 6 Each for $2,098.02 |
|
A867-500
|
Fisher Chemical
FLA867500 |
500g | Glass Bottle |
Each for $627.27
Case of 6 Each for $3,763.62 |
|
Chemical Identifiers
28300-74-5 | |
C8H4K2O12Sb2 · 3 H2O | |
MFCD00167056 | |
Tartrated Antimony |
Specifications
Pass Test | |
2.7% max. | |
Meets Requirements | |
100°C | |
2.0 | |
99.0 to 103.0 % | |
MFCD00167056 | |
667.85 | |
99.0 to 103.0% | |
Antimony Potassium Tartrate Trihydrate |
Pass Test | |
Glass Bottle | |
White | |
250g | |
28300-74-5 | |
C8H4K2O12Sb2 · 3 H2O | |
Tartrated Antimony | |
Solid | |
USP |
Safety and Handling

DOTInformation : DOT Class 6.1, : Poison