missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) telluride, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4592209

 View more versions of this product

Catalog No. AA4592209



Specifications

Specifications

Antimony (III) telluride
(metals basis)
MFCD00016320
antimony telluride, tellanylidenestibanyl tellanyl stibanetellone, antimon iii tellurid, antimony iii telluride, antimony telluride,, antimony iii telluride, beads, antimony iii telluride, powder,-325 mesh trace metals basis
[Sb]=[Te].[Sb]=[Te].[Te]
626.32
626.3
99.999%
−100 Mesh
Gray
10g
1327-50-0
Sb2Te3
UN1549
BPDQXJZWVBPDSN-UHFFFAOYSA-N
tellanylideneantimony;tellurium
6369653
Powder
6.5g/mL
Odorless
629°C

Chemical Identifiers

1327-50-0
626.32
BPDQXJZWVBPDSN-UHFFFAOYSA-N
6369653
[Sb]=[Te].[Sb]=[Te].[Te]
Sb2Te3
MFCD00016320
antimony telluride, tellanylidenestibanyl tellanyl stibanetellone, antimon iii tellurid, antimony iii telluride, antimony telluride,, antimony iii telluride, beads, antimony iii telluride, powder,-325 mesh trace metals basis
tellanylideneantimony;tellurium
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

SDS Documents