missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) sulfide, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1168309

 View more versions of this product

Catalog No. AA1168309



Specifications

Specifications

Antimony(III) sulfide
(metals basis)
UN1549
339.68
99.999%
550°C
1345-04-6
MFCD00011217
14,713
Powder
−60 Mesh
10g

Chemical Identifiers

1345-04-6
MFCD00011217
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

TSCA : TSCA

Documents