Learn More
Antimony(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
Vacuum deposition
Supplier: Thermo Scientific Chemicals 04037130
Description
Antimony Trisulfide is used in the production of pyrotechnics and as a yellow pigment for glass and porcelain. In Vacuum deposition. Used in preparation of Vermilion or yellow pigment, for antimony salts, percussion caps, camouflage paints, and in ruby glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Antimony(III) sulfide | |
1345-04-6 | |
250 g | |
S3Sb2 | |
UN1549 | |
Insoluble in water. Soluble in concentrated acids. | |
[S--].[S--].[S--].[Sb+3].[Sb+3] | |
339.70 | |
99.9% |
550°C | |
Pieces | |
(metals basis) | |
MFCD00011217 | |
14,713 | |
NVWBARWTDVQPJD-UHFFFAOYSA-N | |
diantimony(3+) trisulfanediide | |
339.68 |
Chemical Identifiers
1345-04-6 | |
339.70 | |
NVWBARWTDVQPJD-UHFFFAOYSA-N | |
[S--].[S--].[S--].[Sb+3].[Sb+3] |
S3Sb2 | |
MFCD00011217 | |
diantimony(3+) trisulfanediide |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H312-H315-H319-H335-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber : 215-713-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only