missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony(III) sulfide, 98% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 12521A1

 View more versions of this product

Catalog No. AA12521A1



Specifications

Specifications

Antimony(III) sulfide
98.0% min (Metal basis)
MFCD00011217
Antimony trisulfide
339.68
Odorless
550°C
1345-04-6
Sb2S3
Black powder
Insoluble in water. Soluble in HCl with H2S evolution
Powder
Black
1kg

Chemical Identifiers

1345-04-6
Sb2S3
MFCD00011217
Antimony trisulfide
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H312-H315-H319-H335-H500

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 215-713-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents