missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony(III) sulfide, 98% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1252130

 View more versions of this product

Catalog No. AA1252130



Specifications

Specifications

Antimony(III) sulfide
98.0% min (Metal basis)
MFCD00011217
Antimony trisulfide
Powder
Odorless
250g
1345-04-6
Sb2S3
Black powder
339.68
4.12-4.64g/cm3
Black

Chemical Identifiers

1345-04-6
Sb2S3
MFCD00011217
Antimony trisulfide
Safety and Handling

Safety and Handling

H331-H302-H411-H401

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 215-713-4

Documents