missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Antimony(III) sulfide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 223595000

 View more versions of this product

Catalog No. AC223595000


 

Specifications Description & Specifications

Specifications

≥97.5% (Sb2S3)
339.67
Dark gray to black
Antimony(III) sulfide
S3Sb2
MFCD00011217
1345-04-6
Glass bottle
Crystalline Powder
500g
15, 701
98%
Sb2S3
98%
Antimony trisulfide

Chemical Identifiers

1345-04-6
S3Sb2
339.67
Antimony trisulfide
≥97.5% (Sb2S3)
98%
Sb2S3
MFCD00011217
Antimony(III) sulfide, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 215-713-4

RTECSNumber : CC9450000

TSCA : TSCA