Please login to your online account to display your discounted pricing

Alfa Aesar™ Antimony(III) selenide, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Antimony(III) selenide
CAS 1315-05-5
Formula Weight 480.38
MDL Number MFCD00016319
UN Number UN3077
Merck Index 14,712
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1313006

  SDS
alfa aesar™
1313006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1313014

  SDS
alfa aesar™
1313014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 480.38
Chemical Name or Material Antimony(III) selenide
TSCA TSCA
Particle Size 3 to 12mm
Physical Form Pieces
Assay 99.999%
Sensitivity Moisture Sensitive
Assay Percent Notes (metals basis)
MDL Number MFCD00016319
CAS 1315-05-5
UN Number UN3077
Merck Index 14,712
Melting Point 611°C
Quantity 5g
Solubility Information Very slightly soluble in water
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.